Merck
  • Expression of Amyloid Precursor Protein, Caveolin-1, Alpha-, Beta-, and Gamma-Secretases in Penumbra Cells after Photothrombotic Stroke and Evaluation of Neuroprotective Effect of Secretase and Caveolin-1 Inhibitors.
Expression of Amyloid Precursor Protein, Caveolin-1, Alpha-, Beta-, and Gamma-Secretases in Penumbra Cells after Photothrombotic Stroke and Evaluation of Neuroprotective Effect of Secretase and Caveolin-1 Inhibitors.
Biomedicines (2022-10-28)
Svetlana Sharifulina, Andrey Khaitin, Valeria Guzenko, Yuliya Kalyuzhnaya, Valentina Dzreyan, Alexandr Logvinov, Natalia Dobaeva, Yan Li, Lei Chen, Bin He, Svetlana Demyanenko
ABSTRACT
Our studies reveal changes in the expression of the main participants in the processing of amyloid precursor protein (APP) in neurons and astrocytes after photothrombotic stroke (PTS). Here we show the increase in the level of N- and C-terminal fragments of APP in the cytoplasm of ischemic penumbra cells at 24 h after PTS and their co-immunoprecipitation with caveolin-1. The ADAM10 α-secretase level decreased in the rat brain cortex on the first day after PTS. Levels of γ-secretase complex proteins presenilin-1 and nicastrin were increased in astrocytes, but not in neurons, in the penumbra after PTS. Inhibitory analysis showed that these changes lead to neuronal death and activation of astrocytes in the early recovery period after PTS. The caveolin-1 inhibitor daidzein shifted APP processing towards Aβ synthesis, which caused astroglial activation. γ-secretase inhibitor DAPT down-regulated glial fibrillary acidic protein (GFAP) in astrocytes, prevented mouse cerebral cortex cells from PTS-induced apoptosis, and reduced the infarction volume. Thus, new generation γ-secretase inhibitors may be considered as potential agents for the treatment of stroke.
Sigma-Aldrich
Anti-BACE1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-β-Amyloid antibody, Mouse monoclonal, clone NAB 228, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Caveolin-1 antibody produced in mouse, clone CAV1, tissue culture supernatant
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Gfap antibody produced in mouse, clone S206A-8, purified immunoglobulin
Sigma-Aldrich
Anti-Nicastrin (ab1) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Presenilin1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Batimastat, ≥98% (HPLC)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse