Merck
  • Preventing erosion of X-chromosome inactivation in human embryonic stem cells.
Preventing erosion of X-chromosome inactivation in human embryonic stem cells.
Nature communications (2022-05-07)
Marissa Cloutier, Surinder Kumar, Emily Buttigieg, Laura Keller, Brandon Lee, Aaron Williams, Sandra Mojica-Perez, Indri Erliandri, Andre Monteiro Da Rocha, Kenneth Cadigan, Gary D Smith, Sundeep Kalantry
ABSTRACT
X-chromosome inactivation is a paradigm of epigenetic transcriptional regulation. Female human embryonic stem cells (hESCs) often undergo erosion of X-inactivation upon prolonged culture. Here, we investigate the sources of X-inactivation instability by deriving new primed pluripotent hESC lines. We find that culture media composition dramatically influenced the expression of XIST lncRNA, a key regulator of X-inactivation. hESCs cultured in a defined xenofree medium stably maintained XIST RNA expression and coating, whereas hESCs cultured in the widely used mTeSR1 medium lost XIST RNA expression. We pinpointed lithium chloride in mTeSR1 as a cause of XIST RNA loss. The addition of lithium chloride or inhibitors of GSK-3 proteins that are targeted by lithium to the defined hESC culture medium impeded XIST RNA expression. GSK-3 inhibition in differentiating female mouse embryonic stem cells and epiblast stem cells also resulted in a loss of XIST RNA expression. Together, these data may reconcile observed variations in X-inactivation in hESCs and inform the faithful culture of pluripotent stem cells.
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^6 units/ml
Sigma-Aldrich
Alsterpaullone, A cell-permeable, potent, reversible, and ATP competitive inhibitor of GSK-3β (IC₅₀ = 4 nM) and Cdk1/cyclin B (IC₅₀ = 35 nM).