Merck
  • Protocol for intracellular and extracellular metabolite detection in human embryonic stem cells.
Protocol for intracellular and extracellular metabolite detection in human embryonic stem cells.
STAR protocols (2021-09-02)
Faxiang Xu, Chengcheng Song, Weiwei Liu, Guokai Chen
ABSTRACT
Metabolic homeostasis is critical for cell pluripotency and differentiation in human embryonic stem cells (hESCs). It has been reported that metabolic changes specifically regulate cellular signaling during hESC differentiation. This protocol describes procedures for both cell culture and detection of intracellular and extracellular metabolites in hESCs by liquid chromatography-mass spectrometry. Metabolites in glycolysis, citric acid cycle, pentose phosphate pathway, and other metabolic processes can be detected using this approach. For complete details on the use and execution of this protocol, please refer to Song et al., (2019), Yang et al., (2019), Meng et al., (2018), and Chen et al., (2011b).
Sigma-Aldrich
Albumin human, Cellastim S, recombinant, expressed in rice, lyophilized powder, suitable for cell culture, low endotoxin, ≥96% (SDS-PAGE)
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
2-Phospho-L-ascorbic acid trisodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Supelco
Methyl heptadecanoate, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture