Merck
  • Mitochondria-rough-ER contacts in the liver regulate systemic lipid homeostasis.
Mitochondria-rough-ER contacts in the liver regulate systemic lipid homeostasis.
Cell reports (2021-03-18)
Irene Anastasia, Nicolò Ilacqua, Andrea Raimondi, Philippe Lemieux, Rana Ghandehari-Alavijeh, Guilhem Faure, Sergei L Mekhedov, Kevin J Williams, Federico Caicci, Giorgio Valle, Marta Giacomello, Ariel D Quiroga, Richard Lehner, Michael J Miksis, Katalin Toth, Thomas Q de Aguiar Vallim, Eugene V Koonin, Luca Scorrano, Luca Pellegrini
ABSTRACT
Contacts between organelles create microdomains that play major roles in regulating key intracellular activities and signaling pathways, but whether they also regulate systemic functions remains unknown. Here, we report the ultrastructural organization and dynamics of the inter-organellar contact established by sheets of curved rough endoplasmic reticulum closely wrapped around the mitochondria (wrappER). To elucidate the in vivo function of this contact, mouse liver fractions enriched in wrappER-associated mitochondria are analyzed by transcriptomics, proteomics, and lipidomics. The biochemical signature of the wrappER points to a role in the biogenesis of very-low-density lipoproteins (VLDL). Altering wrappER-mitochondria contacts curtails VLDL secretion and increases hepatic fatty acids, lipid droplets, and neutral lipid content. Conversely, acute liver-specific ablation of Mttp, the most upstream regulator of VLDL biogenesis, recapitulates this hepatic dyslipidemia phenotype and promotes remodeling of the wrappER-mitochondria contact. The discovery that liver wrappER-mitochondria contacts participate in VLDL biology suggests an involvement of inter-organelle contacts in systemic lipid homeostasis.
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Anti-Apolipoprotein B Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Apolipoprotein E Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MTTP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PMP70 antibody, Mouse monoclonal, clone 70-18, purified from hybridoma cell culture
Sigma-Aldrich
Hematoxylin Solution, Gill No. 2
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
DPX Mountant for histology, slide mounting medium