Merck
  • High glucose-induced oxidative stress accelerates myogenesis by altering SUMO reactions.
High glucose-induced oxidative stress accelerates myogenesis by altering SUMO reactions.
Experimental cell research (2020-08-22)
Xiuxiu Liu, Gabriel Heras, Volker M Lauschke, Jia Mi, Geng Tian, Stefano Gastaldello
ABSTRACT
Skeletal muscle preservation is a dynamic process that involves constant repair and regeneration. However, the regenerative capacity of muscle cells declines in hyperglycemia. This study aimed to explore the molecular mechanisms underlying this glucotoxicity during myoblast differentiation. C2C12 cells were exposed to different concentrations of glucose, to recapitulate the development of skeletal muscles in vivo in normo- and hyperglycemic conditions. In high glucose conditions, we found significant increases in levels of total cellular reactive oxygen species (ROS) and a reorganization of SUMO enzyme transcripts and SUMOylated proteins. Furthermore, in anticipation of the ROS-induced damage to newly formed myotubes, we observed acceleration of myogenesis. Interestingly, we found a tight relationship between SUMOylation of the Histone methyltransferase SET7/9 and the maintenance of sarcomeric structures of newly formed myotubes. Finally, treatment with the antioxidant anacardic acid preserved the function and activity of myotubes generated in high-glucose conditions by interfering with both ROS and SUMO pathways. Combined, these results suggest that increased oxidative stress and modulation of SUMO reactions are key mediators of glucotoxicity and inhibition of these perturbations using antioxidants might improve muscle regeneration in hyperglycemia.
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)
Sigma-Aldrich
Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Sodium deoxycholate monohydrate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Sodium deoxycholate monohydrate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (green)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)