Merck
  • Hop (Humulus lupulus)-derived bitter acids as multipotent bioactive compounds.
Hop (Humulus lupulus)-derived bitter acids as multipotent bioactive compounds.
Journal of natural products (2009-05-30)
Marjan Van Cleemput, Ko Cattoor, Karolien De Bosscher, Guy Haegeman, Denis De Keukeleire, Arne Heyerick
ABSTRACT
Hop acids, a family of bitter compounds derived from the hop plant (Humulus lupulus), have been reported to exert a wide range of effects, both in vitro and in vivo. They exhibit potential anticancer activity by inhibiting cell proliferation and angiogenesis, by inducing apoptosis, and by increasing the expression of cytochrome P450 detoxification enzymes. Furthermore, hop bitter acids are effective against inflammatory and metabolic disorders, which makes them challenging candidates for the treatment of diabetes mellitus, cardiovascular diseases, and metabolic syndrome. This review summarizes the current knowledge on hop bitter acids, including both phytochemical aspects, as well as the biological and pharmacological properties of these compounds.
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture