Merck
  • Cisplatin induced neurotoxicity is mediated by Sarm1 and calpain activation.
Cisplatin induced neurotoxicity is mediated by Sarm1 and calpain activation.
Scientific reports (2020-12-16)
Aysel Cetinkaya-Fisgin, Xinghua Luan, Nicole Reed, Ye Eun Jeong, Byoung Chol Oh, Ahmet Hoke
ABSTRACT
Cisplatin is a commonly used chemotherapy agent with significant dose-limiting neurotoxicity resulting in peripheral neuropathy. Although it is postulated that formation of DNA-platinum adducts is responsible for both its cytotoxicity in cancer cells and side effects in neurons, downstream mechanisms that lead to distal axonal degeneration are unknown. Here we show that activation of calpains is required for both neurotoxicity and formation of DNA-platinum adduct formation in neurons but not in cancer cells. Furthermore, we show that neurotoxicity of cisplatin requires activation of Sarm1, a key regulator of Wallerian degeneration, as mice lacking the Sarm1 gene do not develop peripheral neuropathy as evaluated by both behavioral or pathological measures. These findings indicate that Sarm1 and/or specific calpain inhibitors could be developed to prevent cisplatin induced peripheral neuropathy.
Sigma-Aldrich
Anti-Cisplatin DNA Adducts Antibody, clone ICR4, clone 1CR4, from rat
Sigma-Aldrich
Calpastatin Peptide Ac 184-210, ≥95% (HPLC)
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
Calpain-1, Porcine Erythrocytes, Calpain-1, Porcine Erythrocytes, is a native calpain-1. A heterodimeric cysteine proteinase with low Ca2+ requirement (EC₅₀ = 2 µM).