Skip to Content
Merck
All Photos(1)

Key Documents

MABE416

Sigma-Aldrich

Anti-Cisplatin DNA Adducts Antibody, clone ICR4

clone 1CR4, from rat

Synonym(s):

CP9/19, Cisplatin DNA modification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1CR4, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
dot blot: suitable
immunohistochemistry: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

General description

Cisplatin adducts are Guanine-Guanine (GG) DNA intrastrand cross links caused by the chemical compound Cisplatin. Cisplatin is used as a chemotherapy agent that contains platinum. The crosslinking of the DNA triggers apoptosis in the effected cell. Cisplatin is particularly effective in certain cancers such as testicular cancer. Unfortunately like most chemotherapy agents Cisplatin has many side effects and cancer cells typically will develop resistance to the drug after a period of time. Clone ICR4 was formerly known as clone CP9/19.

Specificity

This antibody demonstrates specificity for cisplatin modified DNA.

Immunogen

Cisplatin modified native DNA.

Application

Detect Cisplatin adducts using this rat monoclonal antibody, Anti-Cisplatin DNA Adducts Antibody, clone ICR4 validated for use in Dot Blot, ELISA & IHC.
ELISA Analysis: A representative lot from an independent laboratory detected Cisplatin DNA Adducts in ELISA (Tilby, M. J., et al. (1991). Cancer Res. 51(1):123-129.; Kothandapani, A., et al. (2011). J Biol Chem. 286(16):14564-14574.; Welters, M. J., et al. (1999). Ann Oncol. 10(1):97-103.; Strobeck, M. W., et al. (2000). Proc Natl Acad Sci USA. 97(14):7748-7753.; Kothandapani, A., et al. (2012). Exp Cell Res. 18(16):1973-1986.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Cisplatin DNA Adducts in Head and Neck Squamous Cell Carcinoma tissues. (Welters, M. J., et al. (1999). Ann Oncol. 10(1):97-103.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Dot Blot in untreated and Cisplatin treated DNA from HeLa cell lysates.

Dot Blot Analysis: A 1:1,000 dilution of this antibody detected Cisplatin DNA Adducts in 1.0 and 0.5 µg of Cisplatin treated DNA from HeLa cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Moon Soo Park et al.
Journal of the American Society of Nephrology : JASN, 13(4), 858-865 (2002-03-26)
Cisplatin, a commonly used chemotherapeutic agent, has a major limitation because of its nephrotoxicity. Recent studies have shown that cisplatin causes apoptotic cell death in renal tubule cells, but the underlying molecular mechanisms remain to be elucidated. In this study
Yonggen Zou et al.
Oncology letters, 15(1), 1097-1102 (2018-02-06)
As an important chemotherapeutic agent for the treatment of osteosarcoma, the effectiveness of cisplatin is considered to be due to its unique properties, which allow it to penetrate the cell membrane and form various DNA-platinum adducts, resulting in genetic alterations
And-1 Coordinates with the FANCM Complex to Regulate Fanconi Anemia Signaling and Cisplatin Resistance.
Zhang, et al.
Cancer research, 82, 3249-3262 (2023)
M Harris et al.
British journal of cancer, 108(4), 859-865 (2013-02-07)
Human papilloma virus (HPV) is implicated in >99% of cervical cancers and ∼40% of head and neck squamous cell carcinoma (HNSCC). We previously targeted E6 oncogene with (188)Rhenium-labelled monoclonal antibody (mAb) C1P5 to HPV16 E6 in cervical cancer and HNSCC.
Adrien Joseph et al.
Journal for immunotherapy of cancer, 9(6) (2021-06-25)
Tumors rewire their metabolism to achieve robust anabolism and resistance against therapeutic interventions like cisplatin treatment. For example, a prolonged exposure to cisplatin causes downregulation of pyridoxal kinase (PDXK), the enzyme that generates the active vitamin B6, and upregulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service