Merck
  • Small Extracellular Vesicle Regulation of Mitochondrial Dynamics Reprograms a Hypoxic Tumor Microenvironment.
Small Extracellular Vesicle Regulation of Mitochondrial Dynamics Reprograms a Hypoxic Tumor Microenvironment.
Developmental cell (2020-08-12)
Irene Bertolini, Jagadish C Ghosh, Andrew V Kossenkov, Sudheer Mulugu, Shiv Ram Krishn, Valentina Vaira, Jun Qin, Edward F Plow, Lucia R Languino, Dario C Altieri
ABSTRACT
The crosstalk between tumor cells and the adjacent normal epithelium contributes to cancer progression, but its regulators have remained elusive. Here, we show that breast cancer cells maintained in hypoxia release small extracellular vesicles (sEVs) that activate mitochondrial dynamics, stimulate mitochondrial movements, and promote organelle accumulation at the cortical cytoskeleton in normal mammary epithelial cells. This results in AKT serine/threonine kinase (Akt) activation, membrane focal adhesion turnover, and increased epithelial cell migration. RNA sequencing profiling identified integrin-linked kinase (ILK) as the most upregulated pathway in sEV-treated epithelial cells, and genetic or pharmacologic targeting of ILK reversed mitochondrial reprogramming and suppressed sEV-induced cell movements. In a three-dimensional (3D) model of mammary gland morphogenesis, sEV treatment induced hallmarks of malignant transformation, with deregulated cell death and/or cell proliferation, loss of apical-basal polarity, and appearance of epithelial-to-mesenchymal transition (EMT) markers. Therefore, sEVs released by hypoxic breast cancer cells reprogram mitochondrial dynamics and induce oncogenic changes in a normal mammary epithelium.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT2
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
MISSION® esiRNA, targeting human MFN1
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
MISSION® esiRNA, targeting human ILK
Sigma-Aldrich
ILK inhibitor, Cpd 22, The ILK inhibitor, Cpd 22 controls the biological activity of ILK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.