Merck
  • Assessment of the in vitro cytochrome P450 (CYP) inhibition potential of nafithromycin, a next generation lactone ketolide antibiotic.
Assessment of the in vitro cytochrome P450 (CYP) inhibition potential of nafithromycin, a next generation lactone ketolide antibiotic.
Xenobiotica; the fate of foreign compounds in biological systems (2020-10-21)
Rajesh Chavan, Vineet Zope, Nilesh Chavan, Kiran Patil, Ravindra Yeole, Sachin Bhagwat, Mahesh Patel
ABSTRACT
Nafithromycin is a next generation lactone ketolide antibiotic slated to enter phase III clinical development in India for the treatment of CABP as a shorter 800 mg-OD X3 day therapeutic regimen. Nafithromycin exhibits potent activity against MDR Streptococcus pneumoniae including erythromycin and telithromycin-resistant resistant strains. Older macrolides/ketolides are reported to be potent inhibitors of CYP3A4/5. To facilitate comparative assessment of drug-drug interaction potential, CYP inhibitory activities of nafithromycin was evaluated in comparison with clarithromycin, telithromycin, cethromycin and solithromycin. CYP inhibitory activities were assessed against key CYP isoforms (CYP1A2, 2B6, 2C8, 2C9, 2C19, 2D6 and CYP3A4/5) using human liver microsomes. Additionally, time-dependent inhibition (TDI), metabolism-based inhibition (MBI) and k inact /K I activities were also investigated for CYP3A4/5. Nafithromycin did not inhibit key CYP enzymes and was found to be a weak inhibitor of CYP3A4/5. Comparator antibiotics were found to be potent inhibitors with 2- to 50-fold leftward shifts in CYP3A4/5 IC50 values, while such shift was not noted for nafithromycin. k inact /K I ratio of nafithromycin was 3- to 153-fold lower than comparator drugs, further substantiating its lower affinity for CYP3A4/5. In sum, weaker inhibition and lower k inact /K I ratio for CYP3A4/5, points towards nafithromycin's lower propensities towards clinical drug-drug interactions as compared to other macrolides/ketolides antibiotics.
Sigma-Aldrich
Montelukast sodium hydrate, ≥98% (HPLC)
Sigma-Aldrich
Dextrorphan
Sigma-Aldrich
Quinidine hydrochloride monohydrate, technical grade
Sigma-Aldrich
(2S,3S)-Hydroxybupropion hydrochloride, ≥98% (HPLC)