Skip to Content
Merck
All Photos(1)

Key Documents

UC205

Sigma-Aldrich

Dextrorphan

Synonym(s):

(+)-3-Hydroxy-17-methylmorphinan, 1,3,4,9,10,10a-Hexahydro-6-hydroxy-2H-10,4a-(iminoethano)-11-methylphenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO
CAS Number:
Molecular Weight:
257.37
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

optical activity

[α]/D ≥+54°

color

white to off-white

mp

≥195 °C

solubility

saline: soluble(lit.)

storage temp.

2-8°C

SMILES string

CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34

InChI

1S/C17H23NO/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17/h5-6,11,14,16,19H,2-4,7-10H2,1H3/t14-,16+,17+/m1/s1

InChI key

JAQUASYNZVUNQP-PVAVHDDUSA-N

Gene Information

human ... GRIN2A(2903)
rat ... Grin2a(24409)

Application

CYP2D6 O-Demethyl metabolite of dextromethorphan
Dextrorphan has been studies as an N-methyl-D-aspartate (NMDA) receptor antagonist in Xenopus oocytes and hippocampal neuron cultures.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Dextrorphan is soluble in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ihab R Kamel et al.
Journal of neurosurgical anesthesiology, 20(4), 241-248 (2008-09-25)
Ketamine, an intravenous anesthetic and a major drug of abuse, is a noncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist. Ketamine's enantiomer, S(+)-ketamine, acts stereoselectively on neuronal NMDA receptors. The purpose of this in vitro study was to compare the direct effects of
Jaydeep Yadav et al.
Drug metabolism and disposition: the biological fate of chemicals, 47(7), 732-742 (2019-05-03)
Nonspecific drug partitioning into microsomal membranes must be considered for in vitro-in vivo correlations. This work evaluated the effect of including lipid partitioning in the analysis of complex TDI kinetics with numerical methods. The covariance between lipid partitioning and multiple
Zhijie Shen et al.
American journal of translational research, 8(12), 5545-5556 (2017-01-13)
Many researches have proved functions of anti-oxidation, endothelial protection and pro-angiogenesis efficiency of Shexiang Baoxin Pill (SBP). This study aims to investigate potential for metabolism-based interaction on CYP450s and transporter based interaction on OATP1B1, BRCP and MDR1. Human primary hepatocytes
Chris G Parsons et al.
Methods in molecular biology (Clifton, N.J.), 403, 15-36 (2008-10-02)
Electrophysiological techniques can be used to great effect to help determine the mechanism of action of a compound. However, many factors can compromise the resulting data and their analysis, such as the speed of solution exchange, expression of additional ion
Muhammad Farooq et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(5), 750-757 (2016-03-12)
Although cytochrome P450 (CYP) 2D6 has been widely considered to be noninducible on the basis of human hepatocyte studies, in vivo data suggests that it is inducible by endo- and xenobiotics. Therefore, we investigated if the experimental conditions routinely used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service