Merck
  • The Unfolded Protein Response Mediator PERK Governs Myeloid Cell-Driven Immunosuppression in Tumors through Inhibition of STING Signaling.
The Unfolded Protein Response Mediator PERK Governs Myeloid Cell-Driven Immunosuppression in Tumors through Inhibition of STING Signaling.
Immunity (2020-04-16)
Eslam Mohamed, Rosa A Sierra, Jimena Trillo-Tinoco, Yu Cao, Patrick Innamarato, Kyle K Payne, Alvaro de Mingo Pulido, Jessica Mandula, Shuzhong Zhang, Paul Thevenot, Subir Biswas, Sarah K Abdalla, Tara Lee Costich, Kay Hänggi, Carmen M Anadon, Elsa R Flores, Eric B Haura, Shikhar Mehrotra, Shari Pilon-Thomas, Brian Ruffell, David H Munn, Juan R Cubillos-Ruiz, Jose R Conejo-Garcia, Paulo C Rodriguez
ABSTRACT
The primary mechanisms supporting immunoregulatory polarization of myeloid cells upon infiltration into tumors remain largely unexplored. Elucidation of these signals could enable better strategies to restore protective anti-tumor immunity. Here, we investigated the role of the intrinsic activation of the PKR-like endoplasmic reticulum (ER) kinase (PERK) in the immunoinhibitory actions of tumor-associated myeloid-derived suppressor cells (tumor-MDSCs). PERK signaling increased in tumor-MDSCs, and its deletion transformed MDSCs into myeloid cells that activated CD8+ T cell-mediated immunity against cancer. Tumor-MDSCs lacking PERK exhibited disrupted NRF2-driven antioxidant capacity and impaired mitochondrial respiratory homeostasis. Moreover, reduced NRF2 signaling in PERK-deficient MDSCs elicited cytosolic mitochondrial DNA elevation and, consequently, STING-dependent expression of anti-tumor type I interferon. Reactivation of NRF2 signaling, conditional deletion of STING, or blockade of type I interferon receptor I restored the immunoinhibitory potential of PERK-ablated MDSCs. Our findings demonstrate the pivotal role of PERK in tumor-MDSC functionality and unveil strategies to reprogram immunosuppressive myelopoiesis in tumors to boost cancer immunotherapy.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
D,L-Sulforaphane, An isothiocyanate isolated from broccoli that acts as a potent inducer of phase II detoxifying enzymes in mouse tissues and murine hepatoma cells in culture.
Sigma-Aldrich
PERK Inhibitor I, GSK2606414, GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC₅₀ = 0.4 nM; [ATP] = 5 µM). Targets PERK in its inactive DFG conformation at the ATP-binding region.
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Roche
DNase I, grade II, from bovine pancreas