Merck
  • Human Mast Cell Proteome Reveals Unique Lineage, Putative Functions, and Structural Basis for Cell Ablation.
Human Mast Cell Proteome Reveals Unique Lineage, Putative Functions, and Structural Basis for Cell Ablation.
Immunity (2020-02-13)
Thomas Plum, Xi Wang, Mandy Rettel, Jeroen Krijgsveld, Thorsten B Feyerabend, Hans-Reimer Rodewald
ABSTRACT
Mast cells are rare tissue-resident cells of importance to human allergies. To understand the structural basis of principle mast cell functions, we analyzed the proteome of primary human and mouse mast cells by quantitative mass spectrometry. We identified a mast-cell-specific proteome signature, indicative of a unique lineage, only distantly related to other immune cell types, including innate immune cells. Proteome comparison between human and mouse suggested evolutionary conservation of core mast cell functions. In addition to specific proteases and proteins associated with degranulation and proteoglycan biosynthesis, mast cells expressed proteins potentially involved in interactions with neurons and neurotransmitter metabolism, including cell adhesion molecules, ion channels, and G protein coupled receptors. Toward targeted cell ablation in severe allergic diseases, we used MRGPRX2 for mast cell depletion in human skin biopsies. These proteome analyses suggest a unique role of mast cells in the immune system, probably intertwined with the nervous system.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Sodium cyanoborodeuteride, 97 atom % D, ≥96% (CP)
Sigma-Aldrich
Formaldehyde-13C, d2 solution, 20 wt. % in D2O, ≥99 atom % 13C, ≥98 atom % D
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture