Merck
  • Odorant receptors from Culex quinquefasciatus and Aedes aegypti sensitive to floral compounds.
Odorant receptors from Culex quinquefasciatus and Aedes aegypti sensitive to floral compounds.
Insect biochemistry and molecular biology (2019-08-24)
Fangfang Zeng, Pingxi Xu, Walter S Leal
ABSTRACT
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1, previously identified to be sensitive to plant-derived compounds. We cloned 5 ORs from Culex quinquefasciatus and two ORs from Aedes aegypti, ie, CquiOR2, CquiOR4, CquiOR5, CquiOR84, CquiOR85, AaegOR14, and AaegOR15 and then deorphanized these receptors using the Xenopus oocyte recording system and a large panel of odorants. 2-Phenylethanol, phenethyl formate, and phenethyl propionate were the best ligands for CquiOR4 somewhat resembling the profile of AaegOR15, which gave the strongest responses to phenethyl propionate, phenethyl formate, and acetophenone. In contrast, the best ligands for CquiOR5 were linalool, PMD, and linalool oxide. CquiOR4 was predominantly expressed in antennae of nonblood fed female mosquitoes, with transcript levels significantly reduced after a blood meal. 2-Phenylethanol showed repellency activity comparable to that of DEET at 1%. RNAi experiments suggest that at least in part 2-phenylethanol-elicited repellency is mediated by CquiOR4 activation.
Sigma-Aldrich
Ethyl 3-hydroxyhexanoate, ≥98%, FG
Sigma-Aldrich
trans-2-Hexenoic acid, 99%
Sigma-Aldrich
6-Methyl-5-hepten-2-one, 99%
Sigma-Aldrich
n-Decyl acetate, ≥95%, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
cis-3-Hexenyl acetate, ≥98%, stabilized, FCC, FG
Sigma-Aldrich
trans,trans-Farnesyl acetate, 95%
Sigma-Aldrich
Ethyl 3-hydroxybutyrate, ≥97%, FG
Sigma-Aldrich
trans-2-Heptenal, ≥95%
Sigma-Aldrich
Nonyl acetate, ≥97%, FCC
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Octylamine, 99%
Sigma-Aldrich
Octyl acetate, ≥99%
Sigma-Aldrich
2-Methoxy-4-propylphenol, ≥99%, FG
Sigma-Aldrich
2,3-Dimethylphenol, 98%
Sigma-Aldrich
trans,trans-Farnesol, 96%
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Sigma-Aldrich
trans-2-Nonenal, 97%
Sigma-Aldrich
trans-3-Hexenoic acid, 97%
Sigma-Aldrich
3,5-Dimethylphenol, ≥99%
Sigma-Aldrich
2-Tridecanone, 99%
Sigma-Aldrich
Phenethyl propionate, ≥98%, FG
Sigma-Aldrich
Phenethyl formate, ≥96%, FG
Sigma-Aldrich
m-Cresol, 99%
Sigma-Aldrich
2-Heptanone, 99%
Sigma-Aldrich
Propyl acetate, ≥99.5%
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
(−)-Menthone, 90%
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
3-Octanol, 99%