Merck
  • VAP-SCRN1 interaction regulates dynamic endoplasmic reticulum remodeling and presynaptic function.
VAP-SCRN1 interaction regulates dynamic endoplasmic reticulum remodeling and presynaptic function.
The EMBO journal (2019-08-24)
Feline W Lindhout, Yujie Cao, Josta T Kevenaar, Anna Bodzęta, Riccardo Stucchi, Maria M Boumpoutsari, Eugene A Katrukha, Maarten Altelaar, Harold D MacGillavry, Casper C Hoogenraad
ABSTRACT
In neurons, the continuous and dynamic endoplasmic reticulum (ER) network extends throughout the axon, and its dysfunction causes various axonopathies. However, it remains largely unknown how ER integrity and remodeling modulate presynaptic function in mammalian neurons. Here, we demonstrated that ER membrane receptors VAPA and VAPB are involved in modulating the synaptic vesicle (SV) cycle. VAP interacts with secernin-1 (SCRN1) at the ER membrane via a single FFAT-like motif. Similar to VAP, loss of SCRN1 or SCRN1-VAP interactions resulted in impaired SV cycling. Consistently, SCRN1 or VAP depletion was accompanied by decreased action potential-evoked Ca2+ responses. Additionally, we found that VAP-SCRN1 interactions play an important role in maintaining ER continuity and dynamics, as well as presynaptic Ca2+ homeostasis. Based on these findings, we propose a model where the ER-localized VAP-SCRN1 interactions provide a novel control mechanism to tune ER remodeling and thereby modulate Ca2+ dynamics and SV cycling at presynaptic sites. These data provide new insights into the molecular mechanisms controlling ER structure and dynamics, and highlight the relevance of ER function for SV cycling.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Grace Bio-Labs Press-To-Seal silicone isolator, No PSA, wells, 1 (Round, 13mm diameter, 0.8mm depth)
Sigma-Aldrich
Anti-SCRN1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2