Merck
  • Assessment of AAV Vector Tropisms for Mouse and Human Pluripotent Stem Cell-Derived RPE and Photoreceptor Cells.
Assessment of AAV Vector Tropisms for Mouse and Human Pluripotent Stem Cell-Derived RPE and Photoreceptor Cells.
Human gene therapy (2018-03-28)
Anai Gonzalez-Cordero, Debbie Goh, Kamil Kruczek, Arifa Naeem, Milan Fernando, Sophia-Martha Kleine Holthaus, Matsuki Takaaki, Samuel J I Blackford, Magdalena Kloc, Leticia Agundez, Robert D Sampson, Shyamanga Borooah, Patrick Ovando-Roche, Manjit S Mehat, Emma L West, Alexander J Smith, Rachael A Pearson, Robin R Ali
ABSTRACT
Adeno-associated viral vectors are showing great promise as gene therapy vectors for a wide range of retinal disorders. To date, evaluation of therapeutic approaches has depended almost exclusively on the use of animal models. With recent advances in human stem cell technology, stem cell-derived retina now offers the possibility to assess efficacy in human organoids in vitro. Here we test six adeno-associated virus (AAV) serotypes [AAV2/2, AAV2/9, AAV2/8, AAV2/8T(Y733F), AAV2/5, and ShH10] to determine their efficiency in transducing mouse and human pluripotent stem cell-derived retinal pigment epithelium (RPE) and photoreceptor cells in vitro. All the serotypes tested were capable of transducing RPE and photoreceptor cells in vitro. AAV ShH10 and AAV2/5 are the most efficient vectors at transducing both mouse and human RPE, while AAV2/8 and ShH10 achieved similarly robust transduction of human embryonic stem cell-derived cone photoreceptors. Furthermore, we show that human embryonic stem cell-derived photoreceptors can be used to establish promoter specificity in human cells in vitro. The results of this study will aid capsid selection and vector design for preclinical evaluation of gene therapy approaches, such as gene editing, that require the use of human cells and tissues.
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Opsin antibody produced in mouse, clone RET-P1, ascites fluid
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Microphthalmia (Mi) Antibody, clone C5, clone C5, Chemicon®, from mouse