Merck
Interleukin-17 family and IL-17 receptors.
Cytokine & growth factor reviews (2003-03-26)
T A Moseley, D R Haudenschild, L Rose, A H Reddi
ABSTRACT
Interleukin-17 (IL-17) is a pro-inflammatory cytokine secreted by activated T-cells. Recently discovered related molecules are forming a family of cytokines, the IL-17 family. The prototype member of the family has been designated IL-17A. Due to recent advances in the human genome sequencing and proteomics five additional members have been identified and cloned: IL-17B, IL-17C, IL-17D, IL-17E and IL-17F. The cognate receptors for the IL-17 family identified thus far are: IL-17R, IL-17RH1, IL-17RL (receptor like), IL-17RD and IL-17RE. However, the ligand specificities of many of these receptors have not been established. The IL-17 signaling system is operative in disparate tissues such as articular cartilage, bone, meniscus, brain, hematopoietic tissue, kidney, lung, skin and intestine. Thus, the evolving IL-17 family of ligands and receptors may play an important role in the homeostasis of tissues in health and disease beyond the immune system. This survey reviews the biological actions of IL-17 signaling in cancers, musculoskeletal tissues, the immune system and other tissues.
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-17F from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-17B human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-17D human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-17E human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)