Skip to Content
Merck
Tiamulin VETRANAL®, analytical standard

Tiamulin

Tiamulin VETRANAL®, analytical standard
Synonym(s):
(1S,2R,3S,4S,6R,7S,14R)-6-[(2-Diethylaminoethylthio)acetoxy]-3-hydroxy-2,4,7,14-tetramethyl-4-vinyltricyclo[5,4,3,01.8]tetradecan-9-one
Empirical Formula (Hill Notation):
C28H47NO4S
CAS Number:
Molecular Weight:
493.74
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2