Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Septa, Blue PTFE/white silicone, pkg/100
Products
Product Category
septa (6)
Brand
Supelco (6)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
27515
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 22 mm × 0.060 in., for use with 20, 40, 60 mL vial
Expand
View Pricing
27512
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 13 mm × 0.060 in., for use with 7 mL vial
Expand
View Pricing
27514
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 18 mm × 0.060 in., for use with 22 mL vial
Expand
View Pricing
27511
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 11 mm × 0.060 in., for use with 4 mL vial
Expand
View Pricing
27513
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 16 mm × 0.060 in., for use with 15 mL vial
Expand
View Pricing
27510-U
Septa, Blue PTFE/white silicone, pkg/100,
blue PTFE/white silicone, diam. × thickness 8 mm × 0.060 in., for use with 2 mL vial - standard opening
Expand
View Pricing