Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor for Eppendorf
®
5427R Centrifuge
Products
Product Category
rotors (9)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP5409710001
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-48-11, aerosol-tight, to hold, 48 x 1.5/2.0 mL tubes, lid
Expand
View Pricing
EP5409715003
Rotor for Eppendorf
®
5427R Centrifuge,
S-24-11-AT, aerosol-tight, to hold, 24 x 1.5/2.0 mL tubes, with lid and wind shield
Expand
View Pricing
EP5409712004
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-48-11, with rotor lid, to hold, 48 x 1.5/2.0 mL tubes
Expand
View Pricing
EP5409700006
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-12-17, aerosol tight, to hold, 12 x 5.0 mL Eppendorf Tube®, lid
Expand
View Pricing
EP5409704001
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-24-11-Kit, aerosol tight, Eppendorf
®
5409704001 (Global), lid
Expand
View Pricing
EP5409714007
Rotor for Eppendorf
®
5427R Centrifuge,
F-45-48-PCR, non aerosol tight, to hold, 48 x 0.2 mL PCR tubes (or 6 x PCR strips)
Expand
View Pricing
EP5409702009
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-24-11, aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid
Expand
View Pricing
EP5409706004
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
Expand
View Pricing
EP5409708007
Rotor for Eppendorf
®
5427R Centrifuge,
FA-45-30-11, non aerosol-tight, to hold, 30 x 1.5/2.0 mL tubes, with rotor lid
Expand
View Pricing