Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
radiello
™
Cartridge Adsorbents
Products
Sustainability
Feature
Manufacturer
Agency
Application
Package Size
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
RAD130
radiello
™
Cartridge Adsorbents,
for sampling BTEX and VOCs (CS
2
Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
Expand
View Pricing
RAD170
radiello
™
Cartridge Adsorbents,
for sampling Hydrogen Sulfide (H
2
S), matrix microporous PE coated with zinc acetate, pk of 20
Expand
View Pricing
RAD165
radiello
™
Cartridge Adsorbents,
for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL
®
, pk of 20
Expand
View Pricing
RAD166
radiello
™
Cartridge Adsorbents,
for sampling HF, NO
2
, and SO
2
, matrix microporous PE impregnated with wet TEA, pk of 20
Expand
View Pricing
RAD168
radiello
™
Cartridge Adsorbents,
for sampling Ammonia (NH
3
), matrix microporous PE impregnated with phosphoric acid, pk of 20
Expand
View Pricing
RAD145
radiello
™
Cartridge Adsorbents,
for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
Expand
View Pricing
RAD172
radiello
™
Cartridge Adsorbents,
for sampling Ozone (O
3
), matrix microporous PE tube with 4,4′-dipyridylethylene coated silica, pk of 20
Expand
View Pricing
RAD169
radiello
™
Cartridge Adsorbents,
for sampling HCl, matrix SS net with silica gel (100-400 μm particle size), pk of 20
Expand
View Pricing
RAD147
radiello
™
Cartridge Adsorbents,
for sampling Phenolic Compounds (thermal desorption), matrix SS net (100 mesh, 4.8 mm diam.), Tenax-TA, pk of 20
Expand
View Pricing
RAD141
radiello
™
Cartridge Adsorbents,
pkg of 20 ea
Expand
View Pricing
RAD132
radiello
™
Cartridge Adsorbents,
for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
Expand
View Pricing