Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyethylene (PE) Frit, 20 μm porosity
Products
Product Category
frits (6)
Brand
Supelco (4)
Quality Segment
Application
Package Size
Pore size (µm)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
57183
Polyethylene (PE) Frit, 20 μm porosity,
for use with 20mL SPE Tube, pk of 40
Expand
View Pricing
57181
Polyethylene (PE) Frit, 20 μm porosity,
for use with 6mL SPE Tube, pk of 60
Expand
View Pricing
57180-U
Polyethylene (PE) Frit, 20 μm porosity,
for use with 3mL SPE Tube, pk of 108
Expand
View Pricing
57182-U
Polyethylene (PE) Frit, 20 μm porosity,
for use with 12mL SPE Tube, pk of 40
Expand
View Pricing
57244
Polyethylene (PE) Frit, 20 μm porosity,
for use with 1mL SPE Tube, pk of 216
Expand
View Pricing
57184
Polyethylene (PE) Frit, 20 μm porosity,
for use with 60mL SPE Tube, pk of 32
Expand
View Pricing