Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PEDOT:PSS
Synonym(s):
PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)
Products
Brand
Sigma-Aldrich (7)
Sustainability
Feature
pH Value
Physical Form
Chemical Composition
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
655201
PEDOT:PSS,
high-conductivity grade, 3.0-4.0% aqueous dispersion
Expand
View Pricing
483095
PEDOT:PSS,
conductive grade, 1.3 wt. % aqueous dispersion
Expand
View Pricing
739324
PEDOT:PSS,
high-conductivity grade, 1.5% aqueous dispersion, neutral pH
Expand
View Pricing
560596
PEDOT:PSS,
low-conductivity grade, 2.7 wt. % aqueous dispersion
Expand
View Pricing
900181
PEDOT:PSS,
high-conductivity grade, 0.5-1 wt. % aqueous dispersion
Expand
View Pricing
739332
PEDOT:PSS,
high-conductivity grade, 1.1% aqueous dispersion, surfactant-free
Expand
View Pricing
739316
PEDOT:PSS,
conductive inkjet ink, 0.8% aqueous dispersion
Expand
View Pricing