Skip to Content
Merck
All Photos(2)

Key Documents

739316

Sigma-Aldrich

PEDOT:PSS

greener alternative

conductive inkjet ink, 0.8% aqueous dispersion

Synonym(s):

Orgacon IJ-1005, PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate), 0.8% in H2O, conductive inkjet ink

Quality Level

form

liquid

contains

1-5% Ethanol
5-10% Diethylene glycol

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.8% in H2O

sheet resistance

110 Ω/sq

refractive index

n20/D 1.340

pH

1.5-2.5

viscosity

7-12 cP(22 °C)

density

0.985 g/mL at 25 °C

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT:PSS) is an organic semiconductor wherein conjugated PEDOT is doped with sulfonated PSS, which acts as a counter ion. PEDOT is responsible for the conduction mechanism and the hydrated colloidal solution formed by PSS.
PEDOT:PSS has high electrical conductivity and good oxidation resistance, the properties which make it suitable for electromagnetic shielding and noise suppression. Thus, the polymeric film formed possesses high transparency throughout the visible light spectrum and even in near IR and near UV regions, displaying virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm was observed.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is used in energy conversion and storage, thus has been enhanced for energy efficiency. Click here for more information.

Application

PEDOT:PSS acts as an intrinsically conductive polymer, which can be coated on a variety of substrates and nanoparticles like fullerenes (C60) for the low-cost printing of electronics and optoelectronics based applications. Conductive hydrogels can be prepared by using PEDOT:PSS with polyethylene glycol-diacrylate, which can be potentially used in tissue engineering.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Legal Information

Product of Agfa-Gevaert N.V.
Orgacon is a trademark of Agfa-Gevaert N.V.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrien Pierre et al.
Advanced materials (Deerfield Beach, Fla.), 26(32), 5722-5727 (2014-06-20)
A combination of surface energy-guided blade coating and inkjet printing is used to fabricate an all-printed high performance, high yield, and low variability organic thin film transistor (OTFT) array on a plastic substrate. Functional inks and printing processes were optimized
Mechanically robust, photopatternable conductive hydrogel composites.
Pal RK, et al.
Reactive and Functional Polymers, 120(4), 66-73 (2017)
New Conducting and Semiconducting Polymers for Organic Photovoltaics.
Sapp S and Luebben S
MRS Online Proceedings Library, 1270(4) (2010)
EFFECTIVENESS OF ANNEALING TREATMENT AND POLYMER BLENDS ON IV CHARACTERISTSICS OF POLYMER SOLAR CELL.
Rosa E and Shobih S
Reaktor, 14(4), 261-266 (2014)
The Influence of PEDOT to PSS Ratio on the Optical Properties of PEDOT: PSS Thin Solid Films-Insight from Spectroscopic Ellipsometry.
Bednarski H, et al.
Acta Physica Polonica A, 130(5), 1242-1244 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service