Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnetic Stir Bar
Products
Product Category
stir bars (8)
Manufacturer
Diameter (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
HS120547
Magnetic Stir Bar,
L × diam. 10 mm × 5 mm
Expand
View Pricing
HS120548
Magnetic Stir Bar,
L × diam. 20 mm × 8 mm
Expand
View Pricing
HS120549
Magnetic Stir Bar,
L × diam. 25 mm × 8 mm
Expand
View Pricing
23226
Magnetic Stir Bar,
for use with For 0.1-2.0mL vessels, pkg of 6 ea, PTFE
Expand
View Pricing
HS120552
Magnetic Stir Bar,
L × diam. 35 mm × 9 mm
Expand
View Pricing
HS120551
Magnetic Stir Bar,
L × diam. 30 mm × 8 mm
Expand
View Pricing
23227
Magnetic Stir Bar,
for use with For 3.0-5.0mL vessels, pkg of 6 ea, PTFE
Expand
View Pricing
HS120550
Magnetic Stir Bar,
L × diam. 28 mm × 8 mm
Expand
View Pricing