![Glutaraldehyde solution for in vitro diagnostic use](/deepweb/assets/sigmaaldrich/product/structures/194/500/db30dba8-9c07-4485-a2b8-113363fde2e9/640/db30dba8-9c07-4485-a2b8-113363fde2e9.png)
Glutaraldehyde Solution
![Glutaraldehyde solution for in vitro diagnostic use](/deepweb/assets/sigmaaldrich/product/structures/194/500/db30dba8-9c07-4485-a2b8-113363fde2e9/640/db30dba8-9c07-4485-a2b8-113363fde2e9.png)
Synonym(s):
Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial
Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Products
Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Research Area
Quality Segment
Application
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing