Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GenElute
™
Bacterial Genomic DNA Kits
Synonym(s):
Gen Elute
Products
Product Category
kits (3)
Brand
Sigma-Aldrich (3)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
NA2110
GenElute
™
Bacterial Genomic DNA Kits,
sufficient for 70 purifications
Expand
View Pricing
NA2120
GenElute
™
Bacterial Genomic DNA Kits,
sufficient for 350 purifications
Expand
View Pricing
NA2100
GenElute
™
Bacterial Genomic DNA Kits,
sufficient for 10 purifications
Expand
View Pricing