Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Constantan- resistance alloy
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF16913189
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.2 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF52590786
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.3 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF14148187
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.2 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF20576392
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.50 mm, size 100 × 110 mm, temper as rolled
Expand
View Pricing
GF69988303
Constantan- resistance alloy,
foil, thickness 0.005 mm, Cu55%/Ni45%, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF11790637
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.005 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF30544703
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.005 mm, length 0.2 m, temper as rolled
Expand
View Pricing
GF83438365
Constantan- resistance alloy,
rod, 10 mm diameter, Cu55%/Ni45%, length 200 mm, temper annealed
Expand
View Pricing
GF43430969
Constantan- resistance alloy,
rod, Cu 55%/Ni 45%, 10 mm diameter, length 1000 mm, temper annealed
Expand
View Pricing
GF34314757
Constantan- resistance alloy,
rod, Cu 55%/Ni 45%, 10 mm diameter, length 100 mm, temper annealed
Expand
View Pricing
GF37962737
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.005 mm, length 1 m, temper as rolled
Expand
View Pricing
GF72786325
Constantan- resistance alloy,
foil, Cu55%/Ni45%, thickness 0.005 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF73847649
Constantan- resistance alloy,
foil, Cu55%/Ni45%, thickness 0.3 mm, size 150 × 150 mm, temper as rolled
Expand
View Pricing
GF87542118
Constantan- resistance alloy,
foil, thickness 0.005 mm, Cu55%/Ni45%, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF35151522
Constantan- resistance alloy,
foil, Cu 55%/Ni 45%, thickness 0.005 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF56991538
Constantan- resistance alloy,
rod, Cu 55%/Ni 45%, 10 mm diameter, length 500 mm, temper annealed
Expand
View Pricing
GF96303708
Constantan- resistance alloy,
foil, thickness 0.005 mm, Cu55%/Ni45%, length 0.5 m, temper as rolled
Expand
View Pricing