Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloramine T trihydrate
Synonym(s):
Chloramine T trihydrate,
N
-Chloro-
p
-toluenesulfonamide sodium salt
Linear Formula:
CH
3
C
6
H
4
SO
2
NClNa · 3H
2
O
CAS Number:
7080-50-4
Molecular Weight:
281.69
EC Number:
204-854-7
Products
Product Category
antiseptic/disinfectants (2)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
1.02426
Chloramine T trihydrate,
GR for analysis ACS,Reag. Ph Eur
Expand
View Pricing
8.18705
Chloramine T trihydrate,
for synthesis
Expand
View Pricing
402869
Chloramine T trihydrate,
ACS reagent, 98%
Expand
View Pricing
1.02424
Chloramine T trihydrate,
extra pure
Expand
View Pricing
31224
Chloramine T trihydrate,
reag. Ph. Eur., 98.0-103.0%
Expand
View Pricing
23270
Chloramine T trihydrate,
purum p.a., for the detection of halogens and bromate, ≥98.0% (RT)
Expand
View Pricing