Skip to Content
Merck
All Photos(1)

Key Documents

1.02426

Supelco

Chloramine T trihydrate

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Chloramine T trihydrate, Tosylchloramide sodium, N-Chloro-4-toluenesulfonamide sodium salt, Chlorine bleaching solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClNNaO2S · 3H2O
CAS Number:
Molecular Weight:
281.69
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-854-7
NACRES:
NB.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

potency

935 mg/kg LD50, oral (Rat)

pH

8-10 (20 °C, 50 g/L in H2O)

solubility

150 g/L

bulk density

540‑680 kg/m3

storage temp.

2-8°C

SMILES string

[S](=NCl)(=O)([O-])c1ccc(cc1)C

InChI

1S/C7H8ClNO2S/c1-6-2-4-7(5-3-6)12(10,11)9-8/h2-5H,1H3,(H,9,10,11)/p-1

InChI key

VQWVWLDBJISZHX-UHFFFAOYSA-M

Application


  • Microbiota of laboratory channel catfish skin mucosa and aquaria water exposed to chloramine-T trihydrate.: This study explores the microbiota composition of catfish skin mucosa and aquarium water treated with chloramine-T trihydrate, providing insights into its antimicrobial effects in aquatic environments (Kittel et al., 2024).

  • Synthesis of Pyrroles via Consecutive 6ο-Electrocyclization/Ring-Contraction of Sulfilimines.: This research presents a novel method for synthesizing pyrroles, utilizing chloramine-T trihydrate in the electrocyclization and ring-contraction processes, highlighting its significance in organic synthesis (Haut et al., 2021).

  • Carbon-dot-loaded Co(x)Ni(1-x)Fe(2)O(4); x = 0.9/SiO(2)/TiO(2) nanocomposite with enhanced photocatalytic and antimicrobial potential: An engineered nanocomposite for wastewater treatment.: Demonstrates the enhanced photocatalytic and antimicrobial properties of a nanocomposite containing chloramine-T trihydrate, emphasizing its potential in environmental remediation applications (Abd Elkodous et al., 2020).


Linkage

Replaces: CX0710-1; CX0710-3; CX0710

Analysis Note

Assay (iodometric): 99.0 - 103.0 %
Identity (IR-spectrum): passes test
Appearance of solution: passes test
pH-value (5 %; water): 8.0 - 10.0
Bromide (Br): passes test
Matter insoluble in ethanol: ≤ 1.5 %
ortho compounds: passes test
Corresponds to ACS,Reag. Ph Eur

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service