Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbopack
™
Carbon Adsorbent
EC Number:
215-609-9
Products
Product Category
resins (5)
Brand
Supelco (4)
Physical Form
Product Line
Agency
USP Suitability
Separation Technique
Application
Package Size
Pore Size (Å)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
10258
Carbopack
™
Carbon Adsorbent,
matrix Carbopack
™
C, 80-100 mesh, bottle of 10 g
Expand
View Pricing
10257
Carbopack
™
Carbon Adsorbent,
matrix Carbopack
™
C, 60-80 mesh, bottle of 10 g
Expand
View Pricing
10436
Carbopack
™
Carbon Adsorbent,
matrix Carbopack
™
X, 40-60 mesh, bottle of 10 g
Expand
View Pricing
10437-U
Carbopack
™
Carbon Adsorbent,
matrix Carbopack
™
X, 60-80 mesh, bottle of 10 g
Expand
View Pricing
10439-U
Carbopack
™
Carbon Adsorbent,
matrix Carbopack
™
X, 120-400 mesh, bottle of 50 g
Expand
View Pricing