Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
PCR tubes
Products
Product Category
tubes (11)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
BR781305
BRAND
®
PCR tubes,
capacity 0.2 mL, colorless tube, individual, attached flat cap
Expand
View Pricing
BR781300
BRAND
®
PCR tubes,
capacity 0.2 mL, colorless tube, individual, attached domed cap
Expand
View Pricing
BR781310
BRAND
®
PCR tubes,
capacity 0.5 mL, colorless tube, individual, attached flat cap
Expand
View Pricing
BR781302
BRAND
®
PCR tubes,
capacity 0.2 mL, yellow tube, individual, attached flat cap
Expand
View Pricing
BR781301
BRAND
®
PCR tubes,
capacity 0.2 mL, pink tube, individual, attached flat cap
Expand
View Pricing
BR781304
BRAND
®
PCR tubes,
capacity 0.2 mL, blue tube, individual, attached flat cap
Expand
View Pricing
BR781312
BRAND
®
PCR tubes,
capacity 0.5 mL, yellow tube, individual, attached flat cap
Expand
View Pricing
BR781303
BRAND
®
PCR tubes,
capacity 0.2 mL, green tube, individual, attached flat cap
Expand
View Pricing
BR781313
BRAND
®
PCR tubes,
capacity 0.5 mL, green tube, individual, attached flat cap
Expand
View Pricing
BR781314
BRAND
®
PCR tubes,
capacity 0.5 mL, blue tube, individual, attached flat cap
Expand
View Pricing
BR781311
BRAND
®
PCR tubes,
capacity 0.5 mL, pink tube, individual, attached flat cap
Expand
View Pricing