Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Nicotinamide adenine dinucleotide, reduced disodium salt
Synonym(s):
β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt
Empirical Formula (Hill Notation):
C
21
H
27
N
7
Na
2
O
14
P
2
CAS Number:
606-68-8
Molecular Weight:
709.40
EC Number:
210-123-3
Products
Product Category
nucleotides (6)
Brand
Sigma-Aldrich (6)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
N6785
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 10 mg (per vial)
Expand
View Pricing
N7410
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 2 mg (per vial)
Expand
View Pricing
N0786
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 1 mg (per vial)
Expand
View Pricing
N9410
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 5 mg (per vial)
Expand
View Pricing
N6660
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 0.2 mg (per vial)
Expand
View Pricing
N9535
β-Nicotinamide adenine dinucleotide, reduced disodium salt,
~98%, pkg of 25-30 mg (per vial)
Expand
View Pricing