Skip to Content
Merck
All Photos(1)

Key Documents

N6660

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced disodium salt

~98%, pkg of 0.2 mg (per vial)

Synonym(s):

β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2
CAS Number:
Molecular Weight:
709.40
Beilstein:
5230241
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~98%

Quality Level

form

powder

packaging

pkg of 0.2 mg (per vial)

SMILES string

[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11+,13-,14+,15-,16+,20-,21+;;/m0../s1

InChI key

QRGNQKGQENGQSE-QUWMEQBESA-L

Looking for similar products? Visit Product Comparison Guide

General description

β-Nicotinamide adenine dinucleotide (β-NAD) regulates energy metabolism and immunity. It is a cofactor for mitochondrial deacetylase sirtuin-3 enzyme and modulates inflammasome assembly. β-NAD supresses interleukin-1β levels in monocytic cells in inflammatory syndromes. β-NAD released by neurosecretory cells is a potential neurotransmitter. β-NAD is a vascular mediator in lung endothelial cells and may play a protective role against cytokine mediated inflammation.

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).
β-Nicotinamide adenine dinucleotide, reduced disodium salt has been used:
  • for the standard curve generation in neurotoxity assay of cortical neurons
  • in triosephosphate isomerase assay in jurkat JE6-1 cells
  • in lactate dehydrogenase assay

Biochem/physiol Actions

Electron donor

Reconstitution

Solutions should be prepared fresh and used promptly.

Other Notes

Packaged based on NADH content as determined by UV-Vis.
This is the common form of NADH; do not confuse with α-NADH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Propionic acid secreted from propionibacteria induces NKG2D ligand expression on human-activated T lymphocytes and cancer cells
Andresen L, et al.
Journal of Immunology, jimmunol-j0803014 (2009)
beta-Nicotinamide adenine dinucleotide is an enteric inhibitory neurotransmitter in human and nonhuman primate colons
Hwang SJ, et al.
Gastroenterology, 140(2), 608-617 (2011)
beta-Nicotinamide adenine dinucleotide attenuates lipopolysaccharide-induced inflammatory effects in a murine model of acute lung injury
Umapathy N, et al.
Experimental Lung Research, 38(5), 223-232 (2012)
13C MR Hyperpolarization of Lactate by Using ParaHydrogen and Metabolic Transformation in Vitro
Cavallari E, et al.
Chemistry?A European Journal , 23(5), 1200-1204 (2017)
Deficiency of ATP13A2 leads to lysosomal dysfunction, alpha-synuclein accumulation, and neurotoxicity
Usenovic M, et al.
The Journal of Neuroscience, 32(12), 4240-4246 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service