Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bismuth(III) chloride
Synonym(s):
Bismuth trichloride, Trichlorobismuth, Bismuth(III) chloride
Empirical Formula (Hill Notation):
BiCl
3
CAS Number:
7787-60-2
Molecular Weight:
315.34
EC Number:
232-123-2
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Anhydrous (2)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
224839
Bismuth(III) chloride,
reagent grade, ≥98%
Expand
View Pricing
254142
Bismuth(III) chloride,
99.99% trace metals basis
Expand
View Pricing
450723
Bismuth(III) chloride,
anhydrous, powder, 99.998% trace metals basis
Expand
View Pricing
1.12403
Bismuth(III) chloride,
98+
Expand
View Pricing
470279
Bismuth(III) chloride,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing