Skip to Content
Merck
All Photos(1)

Key Documents

1.12403

Sigma-Aldrich

Bismuth(III) chloride

98+

Synonym(s):

Bismuth(III) chloride, Bismuth trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiCl3
CAS Number:
Molecular Weight:
315.34
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-123-2
NACRES:
NA.22
form:
solid

Quality Level

form

solid

potency

3334 mg/kg LD50, oral (Rat)

bp

447 °C/1013 hPa

mp

230-232 °C

density

4.75 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Bi+3].[Cl-].[Cl-].[Cl-]

InChI

1S/Bi.3ClH/h;3*1H/q+3;;;/p-3

InChI key

JHXKRIRFYBPWGE-UHFFFAOYSA-K

Analysis Note

Assay (complexometric): ≥ 98.0 %
Identity (ICP-OES): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service