Skip to Content
Merck
Biphenyl sodium complex solution technical, 0.7 M±0.2 M in diethylene glycol diethyl ether

Biphenyl sodium complex solution

Biphenyl sodium complex solution technical, 0.7 M±0.2 M in diethylene glycol diethyl ether
Synonym(s):
Sodium biphenyl complex
Linear Formula:
C6H5C6H5 · Na
CAS Number:
Molecular Weight:
177.20

Products

Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Package Size
Reagent Type
Special Grade