Skip to Content
Merck
All Photos(1)

Key Documents

14446

Sigma-Aldrich

Biphenyl sodium complex solution

technical, 0.7 M±0.2 M in diethylene glycol diethyl ether

Synonym(s):

Sodium biphenyl complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5 · Na
CAS Number:
Molecular Weight:
177.20
Beilstein:
3989646
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

liquid

shelf life

limited shelf life

reaction suitability

reagent type: reductant

packaging

pk of 20 × 15 mL

concentration

0.7 M±0.2 M in diethylene glycol diethyl ether

storage temp.

2-8°C

SMILES string

[Na]c1ccc(cc1)-c2ccccc2

InChI

1S/C12H9.Na/c1-3-7-11(8-4-1)12-9-5-2-6-10-12;/h1,3-10H;

InChI key

UUNVZHGYJSUMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biphenyl sodium complex solution is mainly favorable to the reduction of organic halogen compounds and is commonly used as a defluorination reagent in organic synthesis.

Other Notes

Reagent for the quantitative release of covalently bound halogens for analytic determination ; Degradation of pesticides

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Total Organic Fluorine (TOF) in environmental samples using flow-injection and chromatographic methods
Trojanowicz M, et al.
Analytical Methods : Advancing Methods and Applications, 3(5), 1039-1045 (2011)
Sodium biphenyl method for determination of covalently bound fluorine in organic compounds and biological materials.
P Venkateswarlu
Analytical chemistry, 54(7), 1132-1137 (1982-06-01)
M.V. Kennedy et al.
Journal of Environmental Quality, 1, 63-63 (1972)
U. Goedde et al.
WLB, Wasser, Luft Boden, 35, 75-75 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service