
6-Methylflavone

Empirical Formula (Hill Notation):
C16H12O2
CAS Number:
Molecular Weight:
236.27
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing