SML3158
6-Methylflavone
≥98% (HPLC)
Synonym(s):
6-Methyl-2-phenyl-4H-1-benzopyran-4-one, 6-Methyl-2-phenyl-4H-chromen-4-one
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
mp
119-122 °C (dec.) (lit.)
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
SMILES string
Cc1ccc2OC(=CC(=O)c2c1)c3ccccc3
InChI
1S/C16H12O2/c1-11-7-8-15-13(9-11)14(17)10-16(18-15)12-5-3-2-4-6-12/h2-10H,1H3
InChI key
NOQJBXPAMJLUSS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service