![2,3,4,5,6-Pentafluorobenzyl bromide for GC derivatization, LiChropur™, ≥98.5% (GC)](/deepweb/assets/sigmaaldrich/product/structures/348/986/102a46b0-8fd2-4f73-b822-09838694a449/640/102a46b0-8fd2-4f73-b822-09838694a449.png)
2,3,4,5,6-Pentafluorobenzyl bromide
![2,3,4,5,6-Pentafluorobenzyl bromide for GC derivatization, LiChropur™, ≥98.5% (GC)](/deepweb/assets/sigmaaldrich/product/structures/348/986/102a46b0-8fd2-4f73-b822-09838694a449/640/102a46b0-8fd2-4f73-b822-09838694a449.png)
Synonym(s):
α-Bromo-2,3,4,5,6-pentafluorotoluene, (Bromomethyl)pentafluorobenzene, 2,3,4,5,6-Pentafluorobenzylbromide
Linear Formula:
C6F5CH2Br
CAS Number:
Molecular Weight:
260.99
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-3 of 3