Skip to Content
Merck
2,2,2-Trifluoroethylamine hydrochloride 98%

2,2,2-Trifluoroethylamine hydrochloride

2,2,2-Trifluoroethylamine hydrochloride 98%
Synonym(s):
2-Amino-1,1,1-trifluoroethane hydrochloride
Linear Formula:
CF3CH2NH2 · HCl
CAS Number:
Molecular Weight:
135.52
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing