Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Dodecanol
Synonym(s):
Alcohol C
12
, Dodecyl alcohol, Lauryl alcohol, 1-Dodecanol
Linear Formula:
CH
3
(CH
2
)
11
OH
CAS Number:
112-53-8
Molecular Weight:
186.33
EC Number:
203-982-0
Products
Product Category
solvents (2)
Brand
Sigma-Aldrich (2)
Supelco (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
ACS reagent (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
126799
1-Dodecanol,
reagent grade, 98%
Expand
View Pricing
443816
1-Dodecanol,
ACS reagent, ≥98.0%
Expand
View Pricing
8.03462
1-Dodecanol,
for synthesis
Expand
View Pricing
75544
1-Dodecanol,
analytical standard
Expand
View Pricing
44095
1-Dodecanol,
Selectophore
™
, ≥98.0%
Expand
View Pricing