Skip to Content
Merck
All Photos(2)

Key Documents

443816

Sigma-Aldrich

1-Dodecanol

ACS reagent, ≥98.0%

Synonym(s):

Alcohol C12, Dodecyl alcohol, Lauryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
Beilstein:
1738860
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39020229
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥98.0%
bp:
260-262 °C (lit.)
vapor pressure:
0.1 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

7.4 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

≥98.0%

form

solid or liquid

autoignition temp.

500 °F

expl. lim.

4 %

impurities

≤0.05% water

color

APHA: ≤10

refractive index

n20/D 1.442 (lit.)

bp

260-262 °C (lit.)

mp

22-26 °C (lit.)

solubility

water: slightly soluble 1 g/L at 23 °C

density

0.833 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCO

InChI

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Dodecanol is a long chain alkyl alcohol.[1] Its density and viscosity over a temperature range, 303.15 to 323.15K have been determined.[2] The thermodynamics of the interaction between 1-dodecanol and sodium dodecyl sulfate (SDS) micellar solutions has been reported.[3] This solvent meets ACS specifications and can be used for processes requiring strict quality conditions such as analytical testing.

Application

1-Dodecanol may be used in the synthesis of the following:
  • Monododecyl glyceryl ethers via etherification by reacting with glycerol in the presence of 10mol% of 1-bromododecane.[1]
  • ω- and (ω-1)-Hydroxy derivatives via hydroxylation reaction catalyzed by frog liver microsomes.[4]
  • Dodecanal via oxidation in the presence of sodium bromate and ruthenium catalyst.[5]
It may be used to extract polycyclic aromatic hydrocarbons (PAHs) from water samples via dispersive liquid-liquid microextraction technique.[6]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homogeneously-catalyzed etherification of glycerol with 1-dodecanol.
Gaudin P, et al.
Catalysis Science & Technology, 1(4), 616-620 (2011)
Cerium or ruthenium catalyzed oxidation of alcohols to carbonyl compounds by means of sodium bromate.
Kanemoto S, et al.
Bulletin of the Chemical Society of Japan, 59(1), 105-108 (1986)
Y Miura
Lipids, 16(10), 721-725 (1981-10-01)
Frog liver microsomes catalyzed the hydroxylation of 1-dodecanol into the corresponding omega- and (omega-1)-hydroxy derivatives. The hydroxylation rate for 1-dodecanol was much lower than that for lauric acid. Both NADPH and O2 were required for hydroxylation activity. NADH had no
Density and viscosity of 1-alkanols.
Saleh MA, et al.
Physics and Chemistry of Liquids, 42(6), 615-623 (2004)
Thermodynamic studies of 1-dodecanol and SDS in aqueous solutions.
Abe M and Ogino K.
Journal of Colloid and Interface Science, 80(1), 146-152 (1981)

Questions

  1. The 1-dodecanol comes in solid form but I need it in solution. Which solvent can I use to do so?

    1 answer
    1. 1-dodecanol is not offered in liquid format only. This product , along with product 126799 and W261718, consists of a "Solid or Liquid" specification. The melting point of the product (mp 22-26 °C (lit.) is around room temperature - the ambient temperature will influence whether or not this product will be in its solid or liquid form. That is why the specification lists the form as either solid or liquid.

      In order to ensure you receive a lot with the "Liquid" specification result, please contact Technical Service as they can view the batch material in stock and see the appearance specification results per batch. Please navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquiries" under the Products Section with all the required information so that a member of Technical Service can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service