Skip to Content
Merck
All Photos(1)

Key Documents

1609680

USP

Salmeterol Related Compound H

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Hydroxy-4-[2-hydroxy-5-(1-hydroxy-2-{[6-(4-phenylbutoxy)hexyl]amino}ethyl)benzyl]-2-naphthoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H43NO6
CAS Number:
Molecular Weight:
585.73
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salmeterol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N(CC(O)c2cc(c(cc2)O)Cc3c4c(c(c(c3)C(=O)O)O)cccc4)CCCCCCOCCCCc1ccccc1

InChI

1S/C36H43NO6/c38-33-18-17-27(22-29(33)23-28-24-32(36(41)42)35(40)31-16-7-6-15-30(28)31)34(39)25-37-19-9-1-2-10-20-43-21-11-8-14-26-12-4-3-5-13-26/h3-7,12-13,15-18,22,24,34,37-40H,1-2,8-11,14,19-21,23,25H2,(H,41,42)

InChI key

RQBBDIIWJKBGAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salmeterol Related Compound H USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluticasone Propionate and Salmeterol Inhalation Powder
  • Salmeterol Inhalation Powder

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluticasone Propionate and Salmeterol Inhalation Powder
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(2), 2010-2010 (2022)
Salmeterol Inhalation Powder
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(2), 3973-3973 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service