Skip to Content
Merck
All Photos(1)

Documents

1609013

USP

Salicylic acid Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
970950
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salicylic acid

manufacturer/tradename

USP

mp

213-217 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)c1ccc(O)cc1

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

FJKROLUGYXJWQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salicylic acid Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Sodium Salicylate
  • Salicylic Acid
  • Magnesium Salicylate
  • Sodium Salicylate Tablets
  • Magnesium Salicylate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Salicylate Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(4), 4094-4094 (2018)
Magnesium Salicylate Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(4), 2724-2724 (2015)
Sodium Salicylate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(3), 4093-4093 (2020)
Salicylic Acid
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(6), 3970-3970 (2020)
Magnesium Salicylate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(1), 2723-2723 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service