Skip to Content
Merck
All Photos(1)

Key Documents

1544508

USP

Piroxicam

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

piroxicam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)

InChI key

QYSPLQLAKJAUJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piroxicam USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Piroxicam Capsules
  • Piroxicam Compounded Cream
  • Piroxicam Compounded Oral Suspension

Biochem/physiol Actions

Cyclooxygenase inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Lister et al.
European journal of rheumatology and inflammation, 12(4), 6-11 (1993-01-01)
Piroxicam-beta-cyclodextrin is a novel NSAID; it is a supermolecular inclusion complex designed to improve the risk:benefit ratio of piroxicam. In animal studies it has been shown to be as effective as piroxicam as an anti-inflammatory and analgesic agent but with
P Pfitzenmeyer et al.
The Journal of rheumatology, 21(8), 1573-1577 (1994-08-01)
We describe 2 patients who developed pulmonary infiltrates while being treated with piroxicam. There was no satisfactory explanation other than drug induced lung disease for their illness. They had no exposure to pneumotoxic drugs or aerocontaminants. Microorganisms were not cultured
C R Lee et al.
Drugs, 48(6), 907-929 (1994-12-01)
Piroxicam-beta-cyclodextrin is a complex of the established nonsteroidal antiinflammatory drug (NSAID) piroxicam and an inert cyclic macromolecule, beta-cyclodextrin. In clinical trials in patients with rheumatic diseases or pain arising from other conditions, it was as effective an analgesic as standard
R N Brogden et al.
Drugs, 28(4), 292-323 (1984-10-01)
Piroxicam is a chemically different non-steroidal anti-inflammatory drug with a long half-life which enables it to be administered once daily. This member of the oxicam series of compounds is now well established in the treatment of rheumatoid arthritis and osteoarthritis
D Gerber
Drug intelligence & clinical pharmacy, 21(9), 707-710 (1987-09-01)
During the approximately five years (1981-86) that piroxicam has been available in South Africa, the Medicines Safety Centre has received 31 reports of adverse reactions associated with this drug. Among these are two reactions not previously recorded in the literature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service