Skip to Content
Merck
All Photos(1)

Documents

1478403

USP

Olopatadine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid hydrochloride, AL-4943A, KW-4679, Opatanol, Patanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO3·HCl
CAS Number:
Molecular Weight:
373.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

olopatadine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2COc3ccc(CC(O)=O)cc13

InChI

1S/C21H23NO3.ClH/c1-22(2)11-5-8-18-17-7-4-3-6-16(17)14-25-20-10-9-15(12-19(18)20)13-21(23)24;/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24);1H/b18-8-;

InChI key

HVRLZEKDTUEKQH-NOILCQHBSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olopatadine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Olopatadine Hydrochloride Ophthalmic Solution

Biochem/physiol Actions

Olopatadine is a histamine H1 receptor antagonist and mast cell stabilizer. The low level of occupancy H1 receptors in the brain explains the low sedation effect of olopatidine. It is believed that olopatidine is a substrate for P-glycoprotein, which limits its brain penetration.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Ohmori et al.
Arzneimittel-Forschung, 54(12), 809-829 (2005-01-14)
Olopatadine hydrochloride (CAS 140462-76-6, KW-4679, AL-4943A; hereinafter referred to as olopatadine) is a novel antiallergic drug that is a selective histamine H1 receptor antagonist possessing inhibitory effects on the release of inflammatory lipid mediators such as leukotriene and thromboxane from
Hiroshi Fujishima et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 113(4), 476-481 (2014-08-29)
Epinastine hydrochloride is a selective histamine H1 receptor antagonist that also inhibits IgE receptor-mediated histamine release from mast cells. To show the superiority of epinastine 0.05% ophthalmic solution (epinastine) to placebo ophthalmic solution (placebo) and noninferiority to olopatadine 0.1% ophthalmic
[Pharmacological profile and clinical efficacy of olopatadine hydrochloride ophthalmic solution (Patanol 0.1% ophthalmic solution)].
Masakazu Watanabe et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 130(3), 221-231 (2007-09-20)
Kenji Ohmori et al.
Japanese journal of pharmacology, 88(4), 379-397 (2002-06-06)
Olopatadine hydrochloride (olopatadine, 11-[(Z)-3-(dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid monohydrochloride) is a novel antiallergic/histamine H1-receptor antagonistic drug that was synthesized and evaluated in our laboratories. Oral administration of olopatadine at doses of 0.03 mg/kg or higher inhibited the symptoms of experimental allergic skin responses
Daisuke Fujisawa et al.
The Journal of allergy and clinical immunology, 134(3), 622-633 (2014-06-24)
Wheal reactions to intradermally injected neuropeptides, such as substance P (SP) and vasoactive intestinal peptide, are significantly larger and longer lasting in patients with chronic urticaria (CU) than in nonatopic control (NC) subjects. Mas-related gene X2 (MrgX2) has been identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service