Skip to Content
Merck
All Photos(1)

Key Documents

1461003

USP

Niacin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Nicotinic acid, 3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

nicotinic acid

manufacturer/tradename

USP

technique(s)

HPLC: suitable

mp

236-239 °C (lit.)

solubility

ethanol: 12.5 g/L at 25 °C
water: soluble 180 g/L at 20 °C

density

1.473 g/cm3 at 25 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Niacin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Niacin Tablets
  • Niacin Injection
  • Niacin Extended-Release Tablets
  • Inositol Niacinate
  • Niacinamide
  • Water-Soluble Vitamins Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oil- and Water-Soluble Vitamins Capsules
United States Pharmacopeia, 40(5), 5389-5389 (2017)
Niacin Extended-Release Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(4), 3141-3141 (2020)
Niacin
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 3138-3138 (2020)
Water-Soluble Vitamins Capsules
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 5499-5499 (2016)
NIACIN OR NIACINAMIDE ASSAY
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(5), 6704-6704 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service