Skip to Content
Merck
All Photos(1)

Key Documents

1443409

USP

Miconazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O
CAS Number:
Molecular Weight:
416.13
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

miconazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)C(OCc3c(cc(cc3)Cl)Cl)C[n]2cncc2

InChI

1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2

InChI key

BYBLEWFAAKGYCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Miconazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Miconazole Compounded Ophthalmic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

C Oliver Morton et al.
BMC microbiology, 14, 261-261 (2014-10-16)
Superficial infections of the skin and mucous membranes caused by dermatophyte fungi are amongst the most common and challenging infections to treat. Previously we demonstrated the phototoxic effects of photodynamic therapy (PDT) towards Trichophyton rubrum, using a green laser to
Roman Holič et al.
Biochimica et biophysica acta, 1842(10), 1483-1490 (2014-07-30)
Pdr16p is considered a factor of clinical azole resistance in fungal pathogens. The most distinct phenotype of yeast cells lacking Pdr16p is their increased susceptibility to azole and morpholine antifungals. Pdr16p (also known as Sfh3p) of Saccharomyces cerevisiae belongs to
M G Bueno et al.
Oral diseases, 21(1), 57-65 (2013-11-14)
The antimicrobial action of five drugs incorporated in temporary denture relines on the fungal biofilm was evaluated. A Candida albicans biofilm (SC5314) was formed on specimens (10 × 1 mm) of materials (Trusoft and Softone) modified or not (control) by
Pénélope A Andreux et al.
Scientific reports, 4, 5285-5285 (2014-06-14)
Mitochondria are semi-autonomous organelles regulated by a complex network of proteins that are vital for many cellular functions. Because mitochondrial modulators can impact many aspects of cellular homeostasis, their identification and validation has proven challenging. It requires the measurement of
Cheshta Sharma et al.
Mycoses, 57(9), 544-552 (2014-04-05)
We investigated the prevalence of vulvovaginal candidiasis due to C. africana in an STD clinic in India and analysed the genetic relatedness of these C. africana isolates with those outside India. A total of 283 germ-tube-positive yeasts were identified by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service